Homophthalic acid, 98%
Homophthalic acid, 98%
Homophthalic acid, 98%
Thermo Scientific Chemicals

Homophthalic acid, 98%

CAS: 89-51-0 | C9H8O4 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 120840250
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.90
Save 6.25
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.90
Save 6.25
Each
Chemical Identifiers
CAS89-51-0
IUPAC Name2-(carboxymethyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyZHQLTKAVLJKSKR-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point178°C to 182°C
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only