Homopiperazine, 98%
Homopiperazine, 98%
Homopiperazine, 98%
Thermo Scientific Chemicals

Homopiperazine, 98%

CAS: 505-66-8 | C5H12N2 | 100.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 120855000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS505-66-8
IUPAC Name1,4-diazepane
Molecular FormulaC5H12N2
InChI KeyFQUYSHZXSKYCSY-UHFFFAOYSA-N
SMILESC1CNCCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<1 %
Appearance (Form)Crystalline mass
Melting point38°C to 42°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only