2-Hydrazinopyridine, 98%
2-Hydrazinopyridine, 98%
2-Hydrazinopyridine, 98%
Thermo Scientific Chemicals

2-Hydrazinopyridine, 98%

CAS: 4930-98-7 | C5H7N3 | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 120870050
Price (USD)
105.65
Online exclusive
117.00
Save 11.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
105.65
Online exclusive
117.00
Save 11.35
Each
Chemical Identifiers
CAS4930-98-7
IUPAC Name2-hydrazinylpyridine
Molecular FormulaC5H7N3
InChI KeyNWELCUKYUCBVKK-UHFFFAOYSA-N
SMILESNNC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystals or crystalline mass
Melting point43°C to 47°C
Appearance (Color)White to orange-brown
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only