PureLink™ RNase A (20 mg/mL)
Invitrogen™

PureLink™ RNase A (20 mg/mL)

PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA. This reagent is also a component자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
1209102110 mL
1209103925 mL
카탈로그 번호 12091021
제품 가격(KRW)
277,000
온라인 행사
Ends: 31-Dec-2026
307,000
할인액 30,000
Each
수량:
10 mL
제품 가격(KRW)
277,000
온라인 행사
Ends: 31-Dec-2026
307,000
할인액 30,000
Each

PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA. This reagent is also a component of the PureLink™ plasmid purification system.

Concentration
20 mg RNase A/mL in 50 mM Tris-HCl (pH 8.0), 10 mM EDTA.

For Research Use Only. Not for use in diagnostic procedures.
사양
포장 유형Bottle
제품라인UltraPure
수량10 mL
배송 조건Room Temperature
농도20 mg/mL
형태Liquid
Unit SizeEach
구성 및 보관
One 10-mL bottle

Store at 2°C to 8°C.

자주 묻는 질문(FAQ)

I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?

Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.

Is RNase A treatment necessary when isolating HMW DNA from cells or tissue samples?

The RNase A treatment is optional but highly recommended for optimal results. RNAse A treatment helps to remove RNA and single stranded DNA, increasing the purity of double stranded DNA. We recommend using the PureLink RNase A (20 mg/mL) (Cat. No. 12091021).

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
Evaluation of proliferation of neural stem cells in vitro and in vivo.
Authors:Morte MI, Carreira BP, Machado V, Carmo A, Nunes-Correia I, Carvalho CM, Araújo IM,
Journal:Curr Protoc Stem Cell Biol
PubMed ID:23404673
This unit describes two basic protocols for the detection of the proliferation of neural stem cells (NSC). The first one addresses cell proliferation in cultures, starting with primary cell cultures isolated from the mouse subventricular zone (SVZ), in which SVZ-derived NSC are kept in culture as neurospheres. By using this ... More