Search
Search

| Catalog Number | Capacity (Metric) | Closure Color | Product Type | Label |
|---|---|---|---|---|
| 120ORG53-1000 | 120 mL | Orange | Pre-assembled Clicktainer Vial | Tabbed |
| 9000-5300 | 90 mL | Closure Not included | Clicktainer Vial | No Label |
| 90GRN53-1000 | 90 mL | Green | Pre-assembled Clicktainer Vial | Tabbed |
| 90ORG53-1000 | 90 mL | Orange | Pre-assembled Clicktainer Vial | Tabbed |
| 90ORG53-1001 | 90 mL | Orange | Clicktainer Vial w/Dispenser Box | Tabbed |
| 90WHT53-1000 | 90 mL | White | Pre-assembled Clicktainer Vial | Tabbed |
| 90WHT53-1001 | 90 mL | White | Pre-assembled Clicktainer Vial | Tabbed, Temperature Strip |
| 90WHT53-2001 | 90 mL | White | Pre-assembled Clicktainer Vial | Tabbed, Temperature Strip |
| 90WHT53-1002 | 90 mL | White | Pre-assembled Clicktainer Vial | Temperature Strip only |
| 90WHT53-2004 | 90 mL | White | Pre-assembled Clicktainer Vial | Temperature Strip Only |
| 90WHT53-2000 | 90 mL | White | Pre-assembled Clicktainer Vial | Non-tabbed |
| 90WHT53-2003 | 90 mL | White | Pre-assembled Clicktainer Vial | No Label |
| 90WHT53-2002 | 90 mL | White | Unassembled Clicktainer Vial | No Label |
| 12000-5300 | 120 mL | Closure Not Included | Clicktainer Vial | No Label |
| 120ORG53-4000 | 120 mL | Orange | Operating Room Specimen Kit | No Label |
| 120WHT53-1000 | 120 mL | White | Pre-assembled Clicktainer Vial | Tabbed |
| 120WHT53-2000 | 120 mL | White | Unassembled Clicktainer Vial | No Label |
| 120WHT53-4000 | 120 mL | White | Operating Room Specimen Kit | No Label |
| 120YEL53-2000 | 120 mL | Yellow | Pre-assembled Clicktainer Vial | Non-tabbed |
| 120WHT53-3000 | 120 mL | White | Midstream Urine Kit | No Label |
| CAPGRN53-1000 | Vial not Included | Green | Cap | No Label |
| CAPORG53-1000 | Vial Not Included | Orange | Cap | No Label |
| CAPWHT53-1000 | Vial Not Included | White | Cap | No Label |
† The term leakproof applies to Thermo Scientific Samco Clicktainer products, 90mL/53mm and 120mL/53mm, that meet the following criteria: a) a vacuum test that draws 95 kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with BS EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by 1 hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco Clicktainer Vials under conditions of their planned applications. Thermo Scientific Samco Clicktainer Vials are leakproof at ambient temperature and pressure when used with their Clicktainer closures.