4-Hydroxybenzoic Acid Hydrazide, 98%
4-Hydroxybenzoic Acid Hydrazide, 98%
4-Hydroxybenzoic Acid Hydrazide, 98%
Thermo Scientific Chemicals

4-Hydroxybenzoic Acid Hydrazide, 98%

CAS: 5351-23-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 121000250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5351-23-5
IUPAC Name4-hydroxybenzohydrazide
Molecular FormulaC7H8N2O2
InChI KeyZMZGIVVRBMFZSG-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only