Specifications
Chemical Name or Material4-Hydroxybenzophenone
Capacity1000 mL
Name Note99+%
CAS1137-42-4
Capacity (Metric)1000 mL
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Melting Point132.0°C to 135.0°C
ColorBrown to White
ColorTransparent
RTECS NumberPC4959775
Diameter (English)4.2 ±0.1 in.
EINECS Number214-507-1
TSCATSCA
For Use With (Application)Storage
Infrared SpectrumAuthentic
Assay Percent Range98% min. (GC)
PackagingGlass bottle
MaterialBorosilicate Glass (3.3)
Molecular FormulaC13H10O2
Linear FormulaHOC6H4COC6H5
MDL NumberMFCD00002355
Quantity100 g
Beilstein08,II,184
Synonym4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
Solubility InformationSolubility in water: insoluble
Quantity6 Pk.
InChI KeyNPFYZDNDJHZQKY-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
IUPAC Name(4-hydroxyphenyl)-phenylmethanone
Molecular Weight (g/mol)198.22
SterilitySterile
PubChem CID14347
ChEBICHEBI:34421
Formula Weight198.22
Percent Purity98+%
Physical FormPowder
Closure IncludedYes
GraduatedNo
MouthNarrow
Height (Metric)19.8 cm
For Use With (Equipment)Chemicals
Height (English)7.8 in.
Chemical LabelCustom Write-on
Closure LinerUnlined
Closure MaterialBorosilicate Glass
Closure Size29/32
Diameter (Metric)107 ±2 mm
Closure TypeStopper
AutoclavableAutoclavable
Capacity (English)33.8 oz.
ShapeRound
Certifications/ComplianceDIN ISO 4796-2
DisposableReusable
Product TypeReagent Bottle
Unit SizeEach