4-Hydroxybenzyl alcohol, 97%
4-Hydroxybenzyl alcohol, 97%
4-Hydroxybenzyl alcohol, 97%
Thermo Scientific Chemicals

4-Hydroxybenzyl alcohol, 97%

CAS: 623-05-2 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 121031000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS623-05-2
IUPAC Name4-(hydroxymethyl)phenol
Molecular FormulaC7H8O2
InChI KeyBVJSUAQZOZWCKN-UHFFFAOYSA-N
SMILESOCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Beige to pink
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only