3-Hydroxybutyric acid, 95%
3-Hydroxybutyric acid, 95%
3-Hydroxybutyric acid, 95%
Thermo Scientific Chemicals

3-Hydroxybutyric acid, 95%

CAS: 300-85-6 | C4H8O3 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 121050050
Price (USD)
175.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
175.00
Each
Chemical Identifiers
CAS300-85-6
IUPAC Name3-hydroxybutanoic acid
Molecular FormulaC4H8O3
InChI KeyWHBMMWSBFZVSSR-UHFFFAOYNA-N
SMILESCC(O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid or liquid
Titration with NaOH94 to 106 %
Infrared spectrumConforms
Appearance (Color)Colorless to white to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only