N-(2-Hydroxyethyl)piperazine, 98.5%
N-(2-Hydroxyethyl)piperazine, 98.5%
N-(2-Hydroxyethyl)piperazine, 98.5%
Thermo Scientific Chemicals

N-(2-Hydroxyethyl)piperazine, 98.5%

CAS: 103-76-4 | C6H14N2O | 130.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
121191000100 gGlass bottle
Catalog number 121191000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-76-4
IUPAC Name2-(piperazin-1-yl)ethan-1-ol
Molecular FormulaC6H14N2O
InChI KeyWFCSWCVEJLETKA-UHFFFAOYSA-N
SMILESOCCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
AppearanceMay discolor to yellow on storage
GC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only