Interested in a bulk or custom version of this product? Request pricing.

MUC1 Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 12123H02H25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Mucin 1 Cell Surface Associated (MUC1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human MUC1 isoform 2 (NP_001018016.1) extracellular domain (Met 1-Gly 167) fused to the Fc region of human IgG1 at the C-terminus.

Activity: Measured by its ability to bind biotinylated mouse ERBB2 in a functional ELISA.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.


Form: Lyophilized
Purity: ≥ 85% by SDS-PAGE
Species: Human
Expression: Mammalian
Gene Symbol: MUC1
Gene Aliases: RP11-263K19.2, CD227, EMA, H23AG, KL-6, MAM6, PEM, PEMT, PUM
Product Size: 5 x 5 µg
Protein Family: CD Proteins
Endotoxin Level: < 1 EU/µg
Protein Subtype: NFκB Pathway Proteins, Intercellular adhesion Molecule
Molecular Weight: 42 kDa
Research Category: Signal Transduction, Oncology
Protein Tag or Fusion: IgG1 Fc Tag
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.


Manuals & protocols