4-Hydroxy-2-mercapto-6-propylpyrimidine, 98%, Thermo Scientific Chemicals
4-Hydroxy-2-mercapto-6-propylpyrimidine, 98%, Thermo Scientific Chemicals
4-Hydroxy-2-mercapto-6-propylpyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-2-mercapto-6-propylpyrimidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
121311000100gPlastic bottle
12131025025 gPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 121311000
Price (EUR)
215,00
Each
Quantity:
100g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
215,00
Each
Chemical Identifiers
CAS51-52-5
IUPAC Name6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Molecular FormulaC7H10N2OS
InChI KeyKNAHARQHSZJURB-UHFFFAOYSA-N
SMILESCCCC1=CC(=O)NC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellowish white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point217°C to 221°C
Titration>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only