3-Hydroxy-3-methyl-2-butanone, 92%
3-Hydroxy-3-methyl-2-butanone, 92%
3-Hydroxy-3-methyl-2-butanone, 92%
Thermo Scientific Chemicals

3-Hydroxy-3-methyl-2-butanone, 92%

CAS: 115-22-0 | C5H10O2 | 102.13 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 121440050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS115-22-0
IUPAC Name3-hydroxy-3-methylbutan-2-one
Molecular FormulaC5H10O2
InChI KeyBNDRWEVUODOUDW-UHFFFAOYSA-N
SMILESCC(=O)C(C)(C)O
さらに表示
Appearance (Color)Clear colorless to yellow to light red
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=90 %
Refractive index1.4140 to 1.4170 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only