4-Hydroxy-4-methyl-2-pentanone, 99%
4-Hydroxy-4-methyl-2-pentanone, 99%
4-Hydroxy-4-methyl-2-pentanone, 99%
4-Hydroxy-4-methyl-2-pentanone, 99%
Thermo Scientific Chemicals

4-Hydroxy-4-methyl-2-pentanone, 99%

CAS: 123-42-2 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 121480100
Price (EUR)
290,00
Each
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
290,00
Each
Add to cart
Chemical Identifiers
CAS123-42-2
IUPAC Name4-hydroxy-4-methylpentan-2-one
Molecular FormulaC6H12O2
InChI KeySWXVUIWOUIDPGS-UHFFFAOYSA-N
SMILESCC(=O)CC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4220 to 1.4250 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only