3-Hydroxy-2-naphthoic acid, 98%
3-Hydroxy-2-naphthoic acid, 98%
3-Hydroxy-2-naphthoic acid, 98%
Acros Organics

3-Hydroxy-2-naphthoic acid, 98%

CAS: 92-70-6 | C11H8O3 | 188.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
121610010
also known as 12161
1 kgPlastic bottle
Catalog number 121610010
also known as 12161
HSN 29182990
Price (INR)
24,569.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
24,569.00
Each
Chemical Identifiers
CAS92-70-6
IUPAC Name3-hydroxynaphthalene-2-carboxylic acid
Molecular FormulaC11H8O3
InChI KeyALKYHXVLJMQRLQ-UHFFFAOYSA-N
SMILESOC(=O)C1=C(O)C=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)Yellow to beige to light brown
Infrared spectrumConforms
Melting point217°C to 223°C (Dec)
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only