4-Hydroxyphenylacetic acid, 98%
4-Hydroxyphenylacetic acid, 98%
4-Hydroxyphenylacetic acid, 98%
Thermo Scientific Chemicals

4-Hydroxyphenylacetic acid, 98%

CAS: 156-38-7 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 121710250
Price (EUR)
62,10
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
62,10
Each
Chemical Identifiers
CAS156-38-7
IUPAC Name2-(4-hydroxyphenyl)acetic acid
Molecular FormulaC8H8O3
InChI KeyXQXPVVBIMDBYFF-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point148°C to 155°C
HPLC>=97.5 %
Appearance (Color)White to cream to light yellow to light brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only