PureLink&trade; <i>Pro</i> 96 total RNA Purification Kit
Actual product may vary
Invitrogen™

PureLink™ Pro 96 total RNA Purification Kit

The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
12173033
also known as 12173-033
2 x 1 LRNA Wash Buffer II (5X)
12173011A384 PrepsRNA Purification Kit
12173022750 mLRNA Lysis Buffer
12173032
also known as 12173-032
5 x 1 LRNA Wash Buffer I
Catalog number 12173033
also known as 12173-033
Price (MXN)
-
Quantity:
2 x 1 L
Product Type:
RNA Wash Buffer II (5X)
The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes. Isolate high yields of total RNA with low genomic DNA contamination from bacteria, yeast, plant, or mammalian samples.

The one-hour procedure requires no handling or disposal of organic waste. RNA binds to a silica membrane and impurities are removed by washing through vacuum filtration or centrifugation. High-purity total RNA is eluted into RNase-free water; ideal for use in a variety of applications, including RT-PCR, qPCR, northern blotting, and nuclease protection assays.

Improved plate design

The PureLink Pro 96 total RNA Purification Kit provides optimized semi-skirted 96-well plates(Figure 3) for improved performance and results. In addition, the plate nozzles with annular collar prevent cross-contamination of samples and improve drying of the silica membrane to prevent ethanol carryover. In addition, sample processing can be performed manually using a benchtop vacuum manifold, by centrifugation (at 2250 x g), or with an automated liquid handling platform. The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5X
For Use With (Application)RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product TypeRNA Wash Buffer II (5X)
Quantity2 x 1 L
Unit SizeEach
Contents & Storage
Store at room temperature.