trans-4-Hydroxy-L-proline, 99+%
trans-4-Hydroxy-L-proline, 99+%
trans-4-Hydroxy-L-proline, 99+%
Thermo Scientific Chemicals

trans-4-Hydroxy-L-proline, 99+%

CAS: 51-35-4 | C5H9NO3 | 131.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 121780250
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.20
Save 13.55
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.20
Save 13.55
Each
Chemical Identifiers
CAS51-35-4
IUPAC Name4-hydroxypyrrolidine-2-carboxylic acid
Molecular FormulaC5H9NO3
InChI KeyPMMYEEVYMWASQN-UHFFFAOYNA-N
SMILESOC1CNC(C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO499.0 to 101.0 %
Sulfated ash=<0.1 %
Ammonium (NH4)=<200 ppm
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only