PureLink™ RNA Micro Scale Kit
Invitrogen™

PureLink™ RNA Micro Scale Kit

The PureLink™ RNA Micro Scale Kit provides rapid purification and concentration of total RNA from a variety of samples (e.g.,Read more
Have Questions?
Catalog NumberQuantity
1218301650 Preps
Catalog number 12183016
Price (USD)
500.65
Online exclusive
534.00
Save 33.35
Each
Quantity:
50 Preps
Price (USD)
500.65
Online exclusive
534.00
Save 33.35
Each
The PureLink™ RNA Micro Scale Kit provides rapid purification and concentration of total RNA from a variety of samples (e.g., LCM tissues, fine needle aspirates, and FACS-sorted cells). RNA purification using the PureLink™ RNA Micro Scale Kit is:

• Effective—expect excellent RNA recovery (up to 100 μg total RNA) from small samples
• Convenient and safe—the extraction procedure uses no phenol/chloroform, CsCl gradients, or precipitation with either LiCl or ethanol
• Fast—obtain high-quality total RNA, typically in just 15 minutes
• Reliable—obtain ready-to-use RNA suitable for any downstream application

A simple and reliable technology for everyday RNA purification
PureLink™ technology combines guanidine-isothiocyanate lysis with the speed, purity, and ease-of-use of silica-membrane purification. The safe and easy procedure can be completed in typically less than 15 minutes with >85% recovery from the first elution. The kit can be used for clean-up of purified RNA from enzymatic reactions, desalting of RNA samples after phenol extraction or ethanol precipitation, and concentration of RNA. Purify RNA from up to 10 mg of tissue or 105 cells. Clean up and concentrate RNA amounts ranging from pg to 100 μg in volumes as small as 10 μL. Purified RNA is ready for use in downstream applications such as RT-PCR, qRT-PCR, northern blots, cDNA library construction, and nuclease protection assays.

A simple and rapid procedure
Homogenize your sample in lysis buffer, add 70% ethanol, and mix. Deposit the resulting solution into a PureLink™ Micro kit column with collection tube, and centrifuge for one minute. Nucleic acids bind to the PureLink™ silica membrane in the column, and after subsequent washes and on-column digestion with DNase, purified RNA can be eluted with RNase-free water.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume12–22 μL
Final Product TypeTotal RNA
For Use With (Application)RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Purification Time15 min. (After sample homogenization)
Quantity50 Preps
ScaleMicro
Shipping ConditionRoom Temperature
Starting Material AmountCells: ≤105
Tissue: ≤5 mg
Enzymatic Reactions: ≤1.2 mL
YieldUp to 17 μg
Isolation TechnologySilica Spin Column
Sample TypeCells, Tissue, Laser Capture Microdissection (LCM), Enzymatic Reactions
Unit SizeEach
Contents & Storage
Box 1 (PureLink™ RNA Micro Kit):
• Lysis Buffer, 125 mL
• Wash Buffer I, 50 mL
• Wash Buffer II, 15 mL
• RNase-free Water, 15.5 mL
• PureLink™ Micro Kit Columns with collection tubes, 50 each
• Collection Tubes, 50 each
• Recovery Tubes, 50 each

Box 2 (PureLink™ DNase/Carrier RNA):
• Lyophilized PureLink™ DNase, 1500 Units
• PureLink™ On-Column DNase 2X Buffer, 1 mL
• RNase-free Water, 1 mL
• Lyophilized PureLink™ Carrier RNA, 300 μg

Store all components of Box 2 at 4°C. Store remaining components at room temperature.

Frequently asked questions (FAQs)

I am using the PureLink RNA Micro Kit, and my RNA yield is very low. Can you offer some tips?

This could be due to incomplete lysis and homogenization. Here are some recommendations:

- Ensure that 10 µL of 2-mercaptoethanol was added per 1 mL of Lysis Buffer.
- Perform all steps at room temperature unless directed otherwise.
- Decrease the amount of starting material used, or increase volume of Lysis Buffer.
- Use the proper homogenization methods according to recommendations in the sample-specific protocols.
- Cut tissue samples into smaller pieces and ensure the tissue is completely immersed in the Lysis Buffer to achieve optimal lysis.

Are the buffers (Lysis Buffer, Wash Buffer I, and Wash Buffer II) used within the PureLink RNA Mini Kit (Cat. No. 12183025) and the PureLink RNA Micro Scale Kit (Cat. No. 12183016) the same?

Yes, the compositions for Lysis Buffer, Wash Buffer I, and Wash Buffer II are the same between the PureLink RNA Mini Kit (Cat. No. 12183025) and the PureLink RNA Micro Scale Kit (Cat. No. 12183016).