PureLink™ RNA Mini Kit, 50 preps - FAQs
PureLink™ RNA Mini Kit, 50 preps - FAQs
View additional product information for PureLink™ RNA Mini Kit - FAQs (A29839, 12183026, 12183025, 12183020, 12183018A)
12 product FAQs found
I've isolated RNA using the PureLink RNA Mini Kit, but see inhibition of downstream enzymatic reactions. What could be causing this?
The presence of ethanol or salt in the purified RNA can inhibit downstream enzymatic reactions. Ensure that you are using the correct order of wash buffers in the kit for washing, and that Wash Buffer II is discarded in the flow-through. Place the spin cartridge into the wash tube and centrifuge the spin cartridge at maximum speed for 2-3 minutes to completely dry the cartridge.
I'm seeing RNA degradation after RNA extraction using the PureLink RNA Mini Kit. What happened?
The RNA could have been contaminated with RNase. Ensure that you are using RNase-free equipment and change gloves frequently. Improper handling can also result in RNA degradation. Ensure samples are processed immediately, and that the lysis is performed quickly after adding the lysis buffer. Lastly, tissues rich in RNase (such as rat pancreas) may require the addition of RNase inhibitors or inactivators to protect the RNA from degradation, or a larger volume of lysis buffer.
I'm getting low RNA yield when using the PureLink RNA Mini Kit. What could be the cause of this and what do you suggest I try?
Low RNA yield can occur due to the following:
- Incomplete lysis and homogenization: ensure that 10 µL of 2-mercaptoethanol was added per milliliter of lysis buffer, perform all steps at room temperature, decrease the amount of starting material used, use proper homogenization methods, and/or cut tissue samples into smaller pieces to ensure complete tissue immersion in the lysis buffer.
- Poor quality starting material: use fresh samples and process immediately after collection.
- Ethanol may not have been added to Wash Buffer II.
- Incorrect elution conditions may have been used: Add RNase-free water and incubate for 1 minute before centrifugation, following the recommendations for elution in the manual. You can also perform a second elution step to recover more RNA.
I'm using the PureLink RNA Mini Kit and my RNA spin cartridge is clogged. What should I do?
Incomplete homogenization or dispersal of precipitate after ethanol addition can lead to clogging of the RNA spin cartridge. Clear the homogenate and remove any particulate or viscous material by centrifugation. Completely disperse any precipitate that forms after adding ethanol to the homogenate. Load only the supernatant onto the RNA spin cartridge to avoid clogging.
What are the differences in technology between the PureLink RNA Mini Kit and RiboPure RNA Purification Kits?
The PureLink RNA Mini Kit provides rapid column-based purification of total RNA, without organic lysis (phenol/chloroform). You can obtain up to 1,000 µg of purified RNA from a single extraction. The RiboPure RNA Purification Kits combine a phenol/guanidine thiocyanate solution with a glass-fiber filter purification method.
Do you have any data showing differences in RNA extraction with TRIzol Reagent, the PureLink RNA Mini Kit, ChargeSwitch Total RNA Cell Kit or the TRIzol Plus RNA Purification Kit?
Please visit our website (http://www.thermofisher.com/content/dam/LifeTech/migration/en/images/ics-organized/applications/nucleic-acid-purification/data-image/560-wide.par.83692.image.559.294.1.gif) for a graph showing purity measurements and RNA integrity number (RIN) comparison of the abovementioned kits.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
I am using the PureLink RNA Mini Kit, but want to perform an on-column DNase I digestion. Do you have suggestions for this?
For on-column digestion, PureLink DNase (Cat. No. 12185010) can be used. Please see the protocol in the appendix of the manual on page 63.
I am using your PureLink RNA Mini Kit. Can I isolate small RNA using this kit?
The PureLink RNA Mini Kit was only validated to recover RNA greater than 200 nt. However, you can try the following modifications to isolate total RNA including small RNA:
- Add more ethanol to the lysate before loading it onto the column. The final ethanol concentration should be at least 55%.
- Increase the ethanol concentration in Wash 1 to at least 55%.
- No on-column DNase treatment can be used without loss of the small RNA.
I'd like to isolate RNA from gram-positive bacteria. What products can you recommend for me? Is a modified lysis protocol needed?
For bacterial samples, you could use our TRIzol Max Bacterial RNA Isolation Kit (Cat. No. 16096040), that contains an extra reagent during homogenization to help with lysis of the bacterial cell walls. You could also use our PureLink RNA Mini Kit (Cat. No. 12183018A), for which you would need to prepare a lyosyme solution to lyse the cell wall.
Have you tested the PureLink RNA Mini Kit (Cat. No. 12183018A) or PureLink miRNA Isolation Kit (Cat. No. K157001) for extraction of RNA with the TaqMan Advanced miRNA Assays?
These kits have not been tested. However, these kits should work with the TaqMan Advanced miRNA Assays. We recommend checking the quality and quantity of small RNAs post-isolation by NanoDrop and Bioanalyzer.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Are the collection tubes for the PureLink RNA Mini Kit (Cat. No. 12183018A) available as a stand-alone item?
Yes, the collection tubes for the PureLink RNA Mini Kit (Cat. No. 12183018A) are available separately as Cat. No. 12282-100. For additional spin cartridges, go through catalog number A29839. However, that one is not listed on the website but it can be ordered by Quick Order.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
Do you offer a kit that will allow me to sequentially isolate gDNA and total RNA from my tissue sample that is not an FFPE (formalin-fixed, paraffin-embedded) sample?
We offer TRIzol reagent that will allow isolation of DNA and RNA from the same sample. Alternatively, we have the following method that has been validated by our R&D team; for sequential isolation of gDNA and total RNA from the same sample. This method involves using 2 of our kits: 1) PureLink RNA Mini Kit (Cat. No. 12183018A, 12183020, 12183025) and 2) PureLink Genomic DNA Mini Kit (Cat. No. K182002, K182000, K182001).
The protocol is detailed below:
Before starting:
- Label all spin columns and buffers from each kit with kit names to prevent confusion.
- Prepare lysis buffer and wash buffers according to the protocol from each kit.
1. Preparing lysates:
- Add 300 µL of lysis buffer (from Purelink RNA Mini Kit, beta-mercaptoethanol added) to cell or tissue sample, lyse the cells as recommended.
2. DNA isolation:
- Load all of the lysate directly onto a Purelink gDNA column (from PureLink Genomic DNA Mini Kit), save flow-through for RNA isolation.
- Centrifuge the Purelink gDNA column at 10,000 x g for 1 min.
- Wash the Purelink gDNA column with 500 µL of Wash Buffer 1 (from PureLink Genomic DNA Mini Kit, ethanol added), centrifuge at 10,000 x g for 1 min.
- Add 500 µL of Wash Buffer 2 (from PureLink Genomic DNA Mini Kit, ethanol added), centrifuge at maximum speed for 3 min to dry the membrane.
- Add 100 µL of Elution Buffer (from PureLink Genomic DNA Mini Kit), incubate at room temperature for 1 min and centrifuge at 10,000 x g for 1 min (yield can be increased if an optional second elution step is added).
- This is purified gDNA.
3. RNA isolation:
- To the above saved flow-through, add same volume of 70% ethanol, mix well and load the lysate/ethanol mix (including all precipitates) onto an RNA spin cartridge (from Purelink RNA Mini Kit).
- Centrifuge at 12,000 x g for 15 sec. Discard flow-through.
- Wash the RNA spin cartridge with 700 µL of Wash Buffer 1 (from Purelink RNA Mini Kit, ethanol added), centrifuge at 12,000 x g for 15 sec.
- Wash twice with 500 µL of Wash Buffer 2 (from Purelink RNA Mini Kit, ethanol added). After the second wash, centrifuge at 12,000 x g for 1 min to dry the membrane.
- Add 50 µL of RNase-free water onto the RNA spin cartridge, incubate at room temperature for 1 min and centrifuge at 12,000 x g for 2 min (yield can be increased if an optional second elution step is added).
- This is purified RNA.