Interested in a bulk or custom version of this product? Request pricing.

NRG4 Recombinant Human Protein

Catalog number: 12183HNCE25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Neuregulin 4 (NRG4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human NRG4 (Q8WWG1) extracellular domain (Pro 2-Phe 62) fused to additional two amino acids (Gly & Pro) at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in 50 mM Tris, 50 mM NaCl, pH 7.2, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Neurobiology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: HRG4
Gene ID: 145957
Gene Symbol: NRG4
Molecular Weight: 6.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Cell Structure⁄Organelle Proteins, Growth Factors & Receptors
Protein Form: Recombinant, Truncated
Protein Name: Neuregulin 1
Protein Subtype: EGF (Epidermal Growth Factor)
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Tag Position: None
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols

Recommended products