8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals

CAS: 148-24-3 | Molecular Formula: C9H7NO | Molecular Weight: 145.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1219000101 kgGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 121900010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS148-24-3
IUPAC Namequinolin-8-ol
Molecular FormulaC9H7NO
InChI KeyMCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILESOC1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.02 %
Appearance (Color)White to light yellow to beige to light brown
Suitability(for Mg determination) Passes test
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

RUO – Research Use Only