8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals

製品番号(カタログ番号) 121905000
価格(JPY)
-
一括またはカスタム形式をリクエストする
化学物質識別子
CAS148-24-3
IUPAC Namequinolin-8-ol
Molecular FormulaC9H7NO
InChI KeyMCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILESOC1=C2N=CC=CC2=CC=C1
さらに表示
Sulfate (SO4)=<0.02 %
Appearance (Color)White to light yellow to beige to light brown
Suitability(for Mg determination) Passes test
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only