4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
Thermo Scientific Chemicals

4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%

CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1219100101 gGlass bottle
Catalog number 121910010
Price (EUR)
34,10
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
34,10
Each
Chemical Identifiers
CAS345909-35-5
IUPAC Name4-hydroxyquinoline-2-carboxylic acid
Molecular FormulaC10H7NO3
InChI KeyHCZHHEIFKROPDY-UHFFFAOYSA-N
SMILESOC(=O)C1=NC2=CC=CC=C2C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration excess NaOH97.5 to 102.5 % (On dry substance)
Water7 to 10 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only