Hypoxanthine, 99.5%
Hypoxanthine, 99.5%
Hypoxanthine, 99.5%
Thermo Scientific Chemicals

Hypoxanthine, 99.5%

CAS: 68-94-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 122010050
Price (USD)
59.90
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
59.90
Each
Chemical Identifiers
CAS68-94-0
IUPAC Name6,7-dihydro-3H-purin-6-one
Molecular FormulaC5H4N4O
InChI KeyFDGQSTZJBFJUBT-UHFFFAOYSA-N
SMILESO=C1N=CNC2=C1NC=N2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<0.5 %
HPLC>=99.4 %
Sulfated ash=<0.2 %
Appearance (Color)White to light yellow to light beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only