1-Indanona, + 99 %, Thermo Scientific Chemicals
1-Indanona, + 99 %, Thermo Scientific Chemicals
1-Indanona, + 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Indanona, + 99 %, Thermo Scientific Chemicals

CAS: 83-33-0 | C9H8O | 132.16 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
10 g
25 g
100 g
Número de catálogo 122110250
Precio (EUR)
82,10
Each
Cantidad:
25 g
Pedido a granel o personalizado
Precio (EUR)
82,10
Each
Identificadores químicos
CAS83-33-0
IUPAC Name2,3-dihydro-1H-inden-1-one
Molecular FormulaC9H8O
InChI KeyQNXSIUBBGPHDDE-UHFFFAOYSA-N
SMILESO=C1CCC2=CC=CC=C12
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Melting point38°C to 42°C
Appearance (Color)Light yellow to brown
Appearance (Form)Crystalline mass
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only