Indole, 99+%
Indole, 99+%
Indole, 99+%
Thermo Scientific Chemicals

Indole, 99+%

CAS: 120-72-9 | C8H7N | 117.15 g/mol
数量:
10 g
25 g
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 122151000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS120-72-9
IUPAC Name1H-indole
Molecular FormulaC8H7N
InChI KeySIKJAQJRHWYJAI-UHFFFAOYSA-N
SMILESN1C=CC2=CC=CC=C12
さらに表示
Appearance (Form)Crystalline powder
Melting point51°C to 53°C
Appearance (Color)White to light pink
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only