Indole-5-carboxylic acid, 98%
Indole-5-carboxylic acid, 98%
Indole-5-carboxylic acid, 98%
Thermo Scientific Chemicals

Indole-5-carboxylic acid, 98%

CAS: 1670-81-1 | C9H6NO2 | 160.15 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 122210050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1670-81-1
IUPAC Name1H-indole-5-carboxylate
Molecular FormulaC9H6NO2
InChI KeyIENZCGNHSIMFJE-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=C2NC=CC2=C1
さらに表示
Titration with NaOH>=97.5 %
Appearance (Color)Yellow to light beige
Infrared spectrumConforms
HPLC>=97.5 %
Melting point207°C to 212°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only