Thermo Scientific Chemicals

Inosine, 99%, Thermo Scientific Chemicals

Catalog number: 122251000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Inosine, 99%, Thermo Scientific Chemicals

Catalog number: 122251000
100 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 122251000
Price (USD)
Quantity
-

Chemical Identifiers

CAS
58-63-9
IUPAC Name
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular Formula
C10H12N4O5
InChI Key
UGQMRVRMYYASKQ-YPLCUDRINA-N
SMILES
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
UV spectrophotometry
98 to 102 % (lambda max. 248.5nm, pH 7)
Sulfated ash
=<0.1 %
Ratio of Absorbances
1.65 to 1.77 (A250/A260)
HPLC
>=98.5 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text