Iodoacetamide, 98%
Iodoacetamide, 98%
Iodoacetamide, 98%
Thermo Scientific Chemicals

Iodoacetamide, 98%

CAS: 144-48-9 | C2H4INO | 184.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 122275000
Price (USD)
2,214.65
Online exclusive
2,460.00
Save 245.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
2,214.65
Online exclusive
2,460.00
Save 245.35
Each
Chemical Identifiers
CAS144-48-9
IUPAC Name2-iodoacetamide
Molecular FormulaC2H4INO
InChI KeyPGLTVOMIXTUURA-UHFFFAOYSA-N
SMILESNC(=O)CI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point>=90°C
Titration Argentometric>=97.5 % (ex I)
Appearance (Color)White to yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only