Iodoacetonitrile, 95%
Iodoacetonitrile, 95%
Iodoacetonitrile, 95%
Thermo Scientific Chemicals

Iodoacetonitrile, 95%

CAS: 624-75-9 | C2H2IN | 166.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 122290050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS624-75-9
IUPAC Name2-iodoacetonitrile
Molecular FormulaC2H2IN
InChI KeyVODKOOOHHCAWFR-UHFFFAOYSA-N
SMILESICC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear red to brown
Appearance (Form)Liquid
GC>=94 %
Infrared spectrumConforms
Refractive index1.5740 to 1.5780 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only