2-Iodoaniline, 98%
2-Iodoaniline, 98%
2-Iodoaniline, 98%
Thermo Scientific Chemicals

2-Iodoaniline, 98%

CAS: 615-43-0 | C6H6IN | 219.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 122300250
Price (USD)
139.65
Special offer
Online exclusive
Ends: 28-Dec-2026
164.00
Save 24.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 28-Dec-2026
164.00
Save 24.35
Each
Chemical Identifiers
CAS615-43-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to brown
Appearance (Form)Powder or crystals or needles
Infrared spectrumConforms
Melting point53°C to 58°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only