(+)-5-Iodo-2'-deoxyuridine, 99%
(+)-5-Iodo-2'-deoxyuridine, 99%
(+)-5-Iodo-2'-deoxyuridine, 99%
Thermo Scientific Chemicals

(+)-5-Iodo-2'-deoxyuridine, 99%

CAS: 54-42-2 | C9H11IN2O5 | 354.10 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 122350050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS54-42-2
IUPAC Name1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11IN2O5
InChI KeyXQFRJNBWHJMXHO-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
さらに表示
Titration with NaOH>=98.5 % (On dried substance)
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Specific optical rotation+28° to +32° (20°C, 589 nm) (c=1, 1 N NaOH)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only