Isatoic anhydride, 98%
Isatoic anhydride, 98%
Isatoic anhydride, 98%
Thermo Scientific Chemicals

Isatoic anhydride, 98%

CAS: 118-48-9 | C8H5NO3 | 163.13 g/mol
Have Questions?
產品號碼QuantityPackaging
122485000500 gPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 122485000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS118-48-9
IUPAC Name2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Molecular FormulaC8H5NO3
InChI KeyTXJUTRJFNRYTHH-UHFFFAOYSA-N
SMILESO=C1NC2=CC=CC=C2C(=O)O1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Beige to brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only