Isobutylamine
Isobutylamine
Isobutylamine
Thermo Scientific Chemicals

Isobutylamine

CAS: 78-81-9 | C4H11N | 73.14 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
產品號碼 122500025
價格 (TWD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS78-81-9
IUPAC Name2-methylpropan-1-amine
Molecular FormulaC4H11N
InChI KeyKDSNLYIMUZNERS-UHFFFAOYSA-N
SMILESCC(C)CN
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to light yellow
Refractive index1.3960 to 1.3980 (20°C, 589 nm)
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only