Isobutyronitrile, 99%
Isobutyronitrile, 99%
Isobutyronitrile, 99%
Thermo Scientific Chemicals

Isobutyronitrile, 99%

CAS: 78-82-0 | C4H7N | 69.11 g/mol
Have Questions?
Catalog NumberQuantity
1225300101 L
Catalog number 122530010
Price (USD)
72.65
Online exclusive
80.20
Save 7.55
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
72.65
Online exclusive
80.20
Save 7.55
Each
Chemical Identifiers
CAS78-82-0
IUPAC Name2-methylpropanenitrile
Molecular FormulaC4H7N
InChI KeyLRDFRRGEGBBSRN-UHFFFAOYSA-N
SMILESCC(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3720 to 1.3740 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only