Isonicotinic acid, 99%
Isonicotinic acid, 99%
Isonicotinic acid, 99%
Thermo Scientific Chemicals

Isonicotinic acid, 99%

CAS: 55-22-1 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 122590025
Price (USD)
234.65
Special offer
Online exclusive
Ends: 28-Dec-2026
276.00
Save 41.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
234.65
Special offer
Online exclusive
Ends: 28-Dec-2026
276.00
Save 41.35
Each
Chemical Identifiers
CAS55-22-1
IUPAC Namepyridine-4-carboxylic acid
Molecular FormulaC6H5NO2
InChI KeyTWBYWOBDOCUKOW-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only