MarrowMAX™ Bone Marrow Medium
Gibco™

MarrowMAX™ Bone Marrow Medium

Fully-supplemented MarrowMAX™ Bone Marrow Medium for short-term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12260001500 mL
12260014100 mL
Catalog number 12260001
Price (USD)
1,288.00
Each
Quantity:
500 mL
Price (USD)
1,288.00
Each

Gibco MarrowMAX Bone Marrow Medium is a fully-supplemented medium developed for the short term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.

MarrowMAX Bone Marrow Medium features:

  • Superior performance
  • Ready-to-use format
  • Quality and performance testing
  • Unique, optimized formulation

Superior performance

MarrowMAX Bone Marrow Medium outperforms other commercially available media containing Giant Cell Tumor (GCT) conditioned media, providing consistent performance with a higher mitotic index and superior chromosomal morphology.

Ready-to-use format

MarrowMAX Bone Marrow Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to 60 days.

Quality and performance testing

Every lot of MarrowMAX Bone Marrow Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Bone marrow cells from the aspirate of a normal adult donor are cultured for 24 hours in MarrowMAX Bone Marrow Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot must pass a sterility test prior to release.

Unique, optimized formulation

MarrowMAX Bone Marrow Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), gentamicin, and L-glutamine. It also contains a novel human stromal cell conditioned medium that consists of a unique blend of hematopoietic growth factors. MarrowMAX Bone Marrow Medium does not contain GCT conditioned medium.

cGMP Manufacturing and Quality System

MarrowMAX Bone Marrow Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications
Cell TypeBone Marrow, Hematopoietic Cell
Concentration1 X
Included AntibioticsGentamicin
Product LineMarrowMAX
Product TypeBone Marrow Medium
Quantity500 mL
Shipping ConditionDry Ice
SpeciesHuman
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red, FBS, Conditioned Medium (from Stromal Cells)
Unit SizeEach
Contents & Storage
Storage conditions: -5°C to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 18 months from date of manufacture. Thawed medium can be stored at 2°C to 8°C for up to 60 days.