Isonicotinic acid hydrazide, 99%
Isonicotinic acid hydrazide, 99%
Isonicotinic acid hydrazide, 99%
Thermo Scientific Chemicals

Isonicotinic acid hydrazide, 99%

CAS: 54-85-3 | C6H7N3O | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 122601000
Price (EUR)
52,65
Online Exclusive
58,70
Save 6,05 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
52,65
Online Exclusive
58,70
Save 6,05 (10%)
Each
Chemical Identifiers
CAS54-85-3
IUPAC Namepyridine-4-carbohydrazide
Molecular FormulaC6H7N3O
InChI KeyQRXWMOHMRWLFEY-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Titration with HClO4>=98.5 % (On dried substance)
Melting point170°C to 174°C
Appearance (Color)Colorless to white to off-white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only