Isonipecotamide, 98%
Isonipecotamide, 98%
Isonipecotamide, 98%
Thermo Scientific Chemicals

Isonipecotamide, 98%

CAS: 39546-32-2 | C6H12N2O | 128.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122611000
Price (EUR)
101,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
101,00
Each
Chemical Identifiers
CAS39546-32-2
IUPAC Namepiperidine-4-carboxamide
Molecular FormulaC6H12N2O
InChI KeyDPBWFNDFMCCGGJ-UHFFFAOYSA-N
SMILESNC(=O)C1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point146°C to 150°C (on dry substance)
Loss on drying=<2 % (70°C)
Appearance (Color)White to light yellow
Water=<2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only