Isoprene, 98%, stabilized
Isoprene, 98%, stabilized
Isoprene, 98%, stabilized
Thermo Scientific Chemicals

Isoprene, 98%, stabilized

CAS: 78-79-5 | C5H8 | 68.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 122670010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-79-5
IUPAC Name2-methylbuta-1,3-diene
Molecular FormulaC5H8
InChI KeyRRHGJUQNOFWUDK-UHFFFAOYSA-N
SMILESCC(=C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4210 to 1.4230 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only