4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%
4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%
4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%
Thermo Scientific Chemicals

4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%

CAS: 83949-32-0 | C12H15NO2·C7H8O3S | 377.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 122920250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS83949-32-0
IUPAC Name4-phenylpiperidin-1-ium-4-carboxylate
Molecular FormulaC12H15NO2
InChI KeyDZZGGKPKWGPNJA-UHFFFAOYSA-N
SMILES[O-]C(=O)C1(CC[NH2+]CC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point225°C to 229°C
Water=<1 % (K.F.)
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 102.5 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only