1-(Diphenylmethyl)piperazine, 97%
1-(Diphenylmethyl)piperazine, 97%
1-(Diphenylmethyl)piperazine, 97%
Thermo Scientific Chemicals

1-(Diphenylmethyl)piperazine, 97%

CAS: 841-77-0 | C17H20N2 | 252.361 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 122931000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS841-77-0
IUPAC Name1-(diphenylmethyl)piperazin-1-ium chloride
Molecular FormulaC17H21ClN2
InChI KeyDLCXUFKAJDKSDD-UHFFFAOYSA-N
SMILES[Cl-].C1C[NH+](CCN1)C(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Form)Powder
Melting point88°C to 93°C
GC>=96.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only