OptiPRO™ SFM
Gibco™

OptiPRO™ SFM

OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12309050100 mL
123090191000 mL
Catalog number 12309050
Price (MXN)
-
Quantity:
100 mL
Customize this product

OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7. Gibco™ OptiPRO™ SFM features:

  • Serum-free, animal origin-free, low protein formulation
  • Direct adaptation for many popular cell lines

Serum-free, animal origin-free, low protein formulation
Gibco™ OptiPRO™ SFM is formulated with a very low protein concentration (< 10 μg/mL), allowing for easier purification of your recombinant protein.

Direct adaptation for many popular cell lines
Gibco™ OptiPRO™ SFM has been designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO and PK-15. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7, allowing you to choose a single medium for multiple cell lines.

cGMP Manufacturing and Quality System
Gibco™ OptiPRO™ SFM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications
Cell LineHeLa, MDCK, BHK-21, PK-15, COS-7, MDBK, VERO
Concentration1 X
Endotoxin LevelLow
FormatBottle(s)
Included AntibioticsNo antibiotics
Product LineGibco
Product TypeSerum Free Medium
Quantity100 mL
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Low Protein, Serum-free
Culture TypeAdherent Cell Culture
FormLiquid
Serum LevelSerum-free
With AdditivesPhenol Red, Sodium Pyruvate, Sodium Bicarbonate, Thymidine, Hypoxanthine
Without AdditivesNo Glutamine
Unit SizeEach
Contents & Storage
Storage conditions: 2 – 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

Frequently asked questions (FAQs)

Do you offer a protocol for titering virus in OptiPRO SFM?

For virus titering in OptiPRO SFM we recommend using Falcon or equivalent 96-well plates and plating cells at a concentration of 1 X 10^6/mL. If you are having trouble growing cells for virus titration in OptiPRO SFM in the 96-well plates, we suggest switching to the EMEM with 5%FBS or DMEM with 5% FBS.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is there a recommended protocol for culturing cell lines in OptiPRO SFM that require sequential adaptation?

The sequential adaptation process is a multi-subculture of cells. We recommend the following passaging sequence:
• First passage: use 25% OptiPRO SFM:75% original stock medium
• Second passage: use 50% OptiPRO SFM:50% original stock medium
• Third passage: use 75% OptiPRO SFM:25% original stock medium
• Fourth and subsequent passages: use 100% OptiPRO SFM

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I add antibiotics to OptiPRO SFM?

You can supplement OptiPRO SFM with 5mL/L Antibiotic-Antimycotic (Cat. No. 15240096)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What concentration of L-glutamine or GlutaMAX-I should I add to OptiPRO SFM before I use it?

We recommend adding L-glutamine or GlutaMAX-I to OptiPRO SFM at a final concentration of 4mM.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.