Acetic acid, sodium salt trihydrate, 99+%, extra pure
Acetic acid, sodium salt trihydrate, 99+%, extra pure
Acetic acid, sodium salt trihydrate, 99+%, extra pure
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99+%, extra pure

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 123240050
Price (EUR)
196,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
196,00
Each
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Chloride (Cl)=<200 ppm
Infrared spectrumConforms
Insoluble matter=<0.05 %
Sulfate (SO4)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only