Upgrade to SuperFi II DNA Polymerase for better performance. List price reduced by 25%.
Platinum™ SuperFi™ DNA Polymerase
Invitrogen™

Platinum™ SuperFi™ DNA Polymerase

Invitrogen Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology for the highest success in PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
12351010100 Reactions
12351050500 Reactions
123512505 x 500 Reactions
Catalog number 12351010
Price (USD)
292.65
Online exclusive
302.00
Save 9.35
Each
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)
292.65
Online exclusive
302.00
Save 9.35
Each
Invitrogen Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology for the highest success in PCR. Featuring >300X Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Benefits of Platinum SuperFi DNA Polymerase

  • Exceptional >300X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult targets including those of sub-optimal purity or high GC content
  • Room temperature reaction setup and 24 hour bench top stability of pre-assembled reactions
  • Amplification of samples with suboptimal purity

Applications

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Long PCR

Notes

  • Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)300X
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
OverhangBlunt
PolymerasePlatinum SuperFi DNA Polymerase
Quantity100 units
Reaction FormatSeparate Components
Shipping ConditionWet or Dry Ice
Size (Final Product)20 kb or less
For Use With (Application)Hot-start PCR, High-fidelity PCR
GC-Rich PCR PerformanceHigh
Reaction SpeedFast
Unit SizeEach
Contents & Storage
• Platinum SuperFi DNA Polymerase (50 μL at 2 U/μL)
• 5X SuperFi Buffer (2 x 1.25 mL)
• 5X SuperFi GC Enhancer (1.25 mL)

Store at –5°C to –30°C.

Frequently asked questions (FAQs)

Can I use Taq polymerase to generate my gene of interest for directional TOPO cloning?

No, your gene of interest must be amplified with a proofreading polymerase such as Platinum SuperFi DNA Polymerase or AccuPrime Pfx DNA Polymerase that leaves blunt ends for directional TOPO cloning.

Which nucleotide analogues can be used with Platinum SuperFi DNA Polymerase?

Platinum SuperFi DNA Polymerase cannot read dUTP-derivatives or dITP in DNA templates, so the use of these analogues is not recommended.Platinum SuperFi DNA Polymerase can incorporate 7-deaza-dGTP and radiolabeled dNTPs. 

I am having difficulties amplifying long targets using Platinum SuperFi DNA Polymerase. What are your recommendations?

Good lab practices are important for long fragment amplification. These include using high-quality templates (pure, fresh and intact) and fresh primer solutions. Optimization steps to consider include decreasing denaturation and extension temperatures, lengthening extension times as recommended in the manual, and increasing template amounts.

Can Platinum SuperFi DNA Polymerase amplify AT-rich targets?

Yes. To improve amplification of AT-rich targets, we recommend reducing the extension temperature to 68 degrees C or add 5-15 mM Tetramethylammonium Chloride (TMAC).

Can Platinum SuperFi DNA Polymerase amplify GC-rich targets?

All Platinum SuperFi product formats are supplied with 5X SuperFi GC Enhancer which is optimized to improve amplification of GC-rich targets (recommended for use with targets containing >65% GC content)