SuperFi™ Buffers
Invitrogen™

SuperFi™ Buffers

Invitrogen SuperFi Buffer is a 5X buffer solution optimized for use with Platinum SuperFi DNA Polymerase.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
12355005Colorless
12356005Green
Catalog number 12355005
Price (BRL)
693,43
Each
Color:
Colorless
Request bulk or custom format
Price (BRL)
693,43
Each
Invitrogen™ SuperFi™ Buffer is a 5X buffer solution optimized for use with Invitrogen™ Platinum™ SuperFi DNA Polymerase. The buffer contains 7.5 mM MgCl2 which provides 1.5 mM MgCl2 in the final reaction.

The SuperFi Green Buffer (Cat. No. 12356005) includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with the enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless SuperFi™ Buffer. (12355005)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorColorless
Includes5X SuperFi Buffer (4 x 1.25 mL)

Store at –5°C to –30°C.
For Use With (Application)Hot-start PCR
Quantity4 x 1.25 mL
Unit SizeEach