Oxytetracycline Dihydrate 95%
Oxytetracycline Dihydrate 95%
Oxytetracycline Dihydrate 95%
Thermo Scientific Chemicals

Oxytetracycline Dihydrate 95%

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
123841000100 gGlass Bottle
12384010010 gGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 123841000
Price (EUR)
657,00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
657,00
Each
Chemical Identifiers
CAS6153-64-6
IUPAC Name(2Z,4S,4aR,5S,5aR,6S,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,6,10,11,12a-pentahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione dihydrate
Molecular FormulaC22H28N2O11
InChI KeyQRTMTZAUGTXZOA-ZGYDFYMOSA-N
SMILESO.O.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)C4=C(O)C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water6 to 9 % (K.F.)
Appearance (Form)Crystalline powder
HPLC94 to 102 % (on anhydrous substance)
Specific optical rotationon anhydrous substance
Appearance (Color)Yellow to beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only