Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.
Thermo Scientific Chemicals

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
6 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 124020010
Price (USD)
56.65
Online exclusive
62.40
Save 5.75
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
56.65
Online exclusive
62.40
Save 5.75
Each
Chemical Identifiers
CAS8042-47-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Oily liquid
Infrared spectrumConforms
Refractive index1.4620 to 1.4730 (20°C, 589 nm)
Density(15°C) 0.830 to 0.865 kg/l
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For IR spectroscopy
RUO – Research Use Only