Dichloromethane, 99+%, extra pure, stabilized with ethanol
Dichloromethane, 99+%, extra pure, stabilized with ethanol
Dichloromethane, 99+%, extra pure, stabilized with ethanol
Thermo Scientific Chemicals

Dichloromethane, 99+%, extra pure, stabilized with ethanol

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 124050010
Price (USD)
50.70
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
50.70
Each
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Residue after evaporation=<0.05 %
Appearance (Form)Liquid
Water=<0.1 % (K.F.)
StabilizerApprox. 0.2 % Ethanol
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only